Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, zak (human) recombinant protein, human zak (np_598407.1, 1 a.a. - 455 a.a.) full-length recombinant prot ein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5667
Supplier:  Abnova
recombinant/native protein, zak (human) recombinant protein, human zak (np_598407.1, 1 a.a. - 455 a.a.) full-length recombinant prot ein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5667
Supplier:  Abnova