Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, wnk3 (human) recombinant protein, human wnk3 (np_065973.2, 1 a.a. - 434 a.a.) partial recombinant protei n with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5664
Supplier:  Abnova
recombinant/native protein, wnk3 (human) recombinant protein, human wnk3 (np_065973.2, 1 a.a. - 434 a.a.) partial recombinant protei n with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5664
Supplier:  Abnova
Related Information