Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, pkn2 (human) recombinant protein, human pkn2 (np_006247.1, 1 a.a. - 984 a.a.) full-length recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5624
Supplier:  Abnova
recombinant/native protein, pkn2 (human) recombinant protein, human pkn2 (np_006247.1, 1 a.a. - 984 a.a.) full-length recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5624
Supplier:  Abnova