Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, lats2 (human) recombinant protein, human lats2 (np_055387.2, 553 a.a. - 1088 a.a.) partial recombinant p rotein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5574
Supplier:  Abnova
recombinant/native protein, lats2 (human) recombinant protein, human lats2 (np_055387.2, 553 a.a. - 1088 a.a.) partial recombinant p rotein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5574
Supplier:  Abnova
Related Information