Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, kit (t670i) (human) recombinant protein, human kit (np_000213.1, 544 a.a. - 976 a.a.) t670i mutant parti al recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Katalog # ABNOP5571
Supplier:  Abnova
recombinant/native protein, kit (t670i) (human) recombinant protein, human kit (np_000213.1, 544 a.a. - 976 a.a.) t670i mutant parti al recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Katalog # ABNOP5571
Supplier:  Abnova
Related Information