Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, kit (d816v) (human) recombinant protein, human kit (np_000213.1, 544 a.a. - 976 a.a.) d816v mutant parti al recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5570
Supplier:  Abnova
recombinant/native protein, kit (d816v) (human) recombinant protein, human kit (np_000213.1, 544 a.a. - 976 a.a.) d816v mutant parti al recombinant protein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5570
Supplier:  Abnova
Related Information