Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, irak1 (human) recombinant protein, human irak1 (np_001560.2, 194 a.a. - 712 a.a.) partial recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5565
Supplier:  Abnova
recombinant/native protein, irak1 (human) recombinant protein, human irak1 (np_001560.2, 194 a.a. - 712 a.a.) partial recombinant pr otein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5565
Supplier:  Abnova