Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, ddr1 (human) recombinant protein, human ddr1 (np_001945.3, 444 a.a. - 876 a.a.) partial recombinant prot ein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5534
Supplier:  Abnova
recombinant/native protein, ddr1 (human) recombinant protein, human ddr1 (np_001945.3, 444 a.a. - 876 a.a.) partial recombinant prot ein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5534
Supplier:  Abnova