Order Entry
Sweden
ContactUsLinkComponent
kit, varicella zoster virus igg elisa kit, varicella zoster virus igg elisa kit is intended for the detection of igg antibody to vzv in human serum or plasma. 1 * 1 KIT
  ABNOKA2262
 :  Abnova
kit, varicella zoster virus igg elisa kit, varicella zoster virus igg elisa kit is intended for the detection of igg antibody to vzv in human serum or plasma. 1 * 1 KIT
  ABNOKA2262
 :  Abnova