Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, trpm7 (human) recombinant protein, human trpm7 (np_060142.3, 1158 a.a. - 186 a.a.) partial recombinant p rotein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5520
Supplier:  Abnova
recombinant/native protein, trpm7 (human) recombinant protein, human trpm7 (np_060142.3, 1158 a.a. - 186 a.a.) partial recombinant p rotein with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Catalog # ABNOP5520
Supplier:  Abnova
Related Information