Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, cdk4/ccnd3 (human) recombinant protein, human cdk4 (np_000066.1, 1 a.a. - 303 a.a.) and ccnd3 (aaa51927. 1, 1 a.a. - 292 a.a.) with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Katalog # ABNOP5514
Supplier:  Abnova
recombinant/native protein, cdk4/ccnd3 (human) recombinant protein, human cdk4 (np_000066.1, 1 a.a. - 303 a.a.) and ccnd3 (aaa51927. 1, 1 a.a. - 292 a.a.) with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Katalog # ABNOP5514
Supplier:  Abnova