Order Entry
Sweden
ContactUsLinkComponent
recombinant/native protein, cdc2l6/ccnc (human) recombinant protein, human cdc2l6 (np_055891.1, 1 a.a. - 502 a.a.) and ccnc (np_0051 81.2, 1 a.a. - 283 a.a.) with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Katalog # ABNOP5509
Supplier:  Abnova
recombinant/native protein, cdc2l6/ccnc (human) recombinant protein, human cdc2l6 (np_055891.1, 1 a.a. - 502 a.a.) and ccnc (np_0051 81.2, 1 a.a. - 283 a.a.) with gst tag expressed in baculovirus infected sf21 cells. 1 * 5 µG
Katalog # ABNOP5509
Supplier:  Abnova