Order Entry
Sweden
ContactUsLinkComponent
recombinant protein, mybphl (human) recombinant protein (p01), human mybphl full-length orf ( adr83131.1, 1 a.a. - 354 a.a.) recombi nant protein with gst-tag at n-terminal. 1 * 25 µG
Catalog # ABNOH00343263-P012
Supplier:  Abnova
recombinant protein, mybphl (human) recombinant protein (p01), human mybphl full-length orf ( adr83131.1, 1 a.a. - 354 a.a.) recombi nant protein with gst-tag at n-terminal. 1 * 25 µG
Catalog # ABNOH00343263-P012
Supplier:  Abnova