Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

AMPSO ≥98%

Glass bottle for solids, 25 g
Catalog # J62378.14
AMPSO ≥98%
Catalog # J62378.14
CAS Number:  68399-79-1
AMPSO ≥98%
AMPSO ≥98%

Glass bottle for solids, 25 g

CAS Number:  68399-79-1
Price
1 250,00 kr
Log in for availability
1 * 25 g
Product Details & Documents

Purity: 98+% CAS No.: 68399-79-1 Molecular Formula: C7H17NO5S Formula Weight: 227.28 Physical Form: Crystals Appearance: White Melting Point: 215-225° Beilstein Reference: 5929916 MDL No.: MFCD00041777

Useful as a buffer

Synonyms:  N-(1,1-Dimethyl-2-hydroxyethyl)-3-amino-2-hydroxypropanesulphonic acid
  • CAS Number:68399-79-1
  • Formula:C₇H₁₇NO₅S
  • MDL Number:MFCD00041777
  • Molecular Weight:227.28 g/mol
  • Melting Pt:215…225 °C
  • Storage Temperature:Ambient
  • EINECS:269-991-7
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.