Buffer solutions, ready to use, referenced at 25 °C, Certipur®
4,01, Potassium hydrogen phthalate, 4 L
: Merck
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
1.09406.4000
: Merck
:
Materials for the precise calibration and monitoring of pH meters.
- Traceable to NIST primary standard and PTB - deviation max. ±0,01 (except pH 10 with ±0,02)
- Larger 4 litre pack sizes in easy to use Titripac® packaging which eliminates contamination issues
- Colour coded buffers at pH values 4,00, 7,00 and 10,00 are available
- Reference temperature: +25 °C
All buffer solutions are measured and qualified in a DIN EN ISO 17025/IEC accredited laboratory for pH measurement.
: Plastic bottles (500 ml) or PE/aluminium Titripac® pack (4 litre)
- Pk:4 L
- pH value:4,01
- Contents:Potassium hydrogen phthalate
- Cat. No.:1.09406.4000
- Environmentally Preferable:Sustainable Packaging



