Buffer solutions, ready to use, referenced at 25 °C, Certipur®
6,00, Citric acid, sodium hydroxide, 500 mL
Leverantör: Merck
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Katalog # 1.99036.0500
Leverantör: Merck
CAS-nummer:
Gå till:
Product Details & Documents
Materials for the precise calibration and monitoring of pH meters.
- Traceable to NIST primary standard and PTB - deviation max. ±0,01 (except pH 10 with ±0,02)
- Larger 4 litre pack sizes in easy to use Titripac® packaging which eliminates contamination issues
- Colour coded buffers at pH values 4,00, 7,00 and 10,00 are available
- Reference temperature: +25 °C
All buffer solutions are measured and qualified in a DIN EN ISO 17025/IEC accredited laboratory for pH measurement.
Packaging: Plastic bottles (500 ml) or PE/aluminium Titripac® pack (4 litre)
Documents
Certificates
Sök efter LOT-nummerCertipur Merck Buffer Calibration Standards for pH Measurements
Product Literature
ProductPage_109407 ProductPage_109408
Product specification
TDS Certified Reference Materials
Datasheet
Specifikationer
Specifikationer
- Pk:500 mL
- pH value:6,00
- Contents:Citric acid, sodium hydroxide
- Cat. No.:1.99036.0500
- Environmentally Preferable:Sustainable Packaging
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



