Order Entry
Sweden
Orders LinkContactUsLinkComponent
Buffer solutions, ready to use, referenced at 25 °C, Certipur®

Buffer solutions, ready to use, referenced at 25 °C, Certipur®

4,00 (red), Citric acid, sodium hydroxide, hydrogen chloride, 500 mL
Catalog # 1.99054.0500
Supplier:   Merck
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Catalog # 1.99054.0500
Supplier:  Merck
CAS Number:  
Buffer solutions, ready to use, referenced at 25 °C, Certipur®
Buffer solutions, ready to use, referenced at 25 °C, Certipur®

4,00 (red), Citric acid, sodium hydroxide, hydrogen chloride, 500 mL

CAS Number:  
Price
625,00 kr
Log in for availability
1 * 500 mL
Product Details & Documents

Materials for the precise calibration and monitoring of pH meters.

  • Traceable to NIST primary standard and PTB - deviation max. ±0,01 (except pH 10 with ±0,02)
  • Larger 4 litre pack sizes in easy to use Titripac® packaging which eliminates contamination issues
  • Colour coded buffers at pH values 4,00, 7,00 and 10,00 are available
  • Reference temperature: +25 °C

All buffer solutions are measured and qualified in a DIN EN ISO 17025/IEC accredited laboratory for pH measurement.

Packaging: Plastic bottles (500 ml) or PE/aluminium Titripac® pack (4 litre)

Specifications

Specifications

  • Pk:
    500 mL
  • pH value:
    4,00 (red)
  • Contents:
    Citric acid, sodium hydroxide, hydrogen chloride

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.