Order Entry
Sweden
Orders LinkContactUsLinkComponent
Risperidone, Reference Standards

Risperidone, Reference Standards

Risperidone System Suitability Mixture, USP reference standard, Vial, 10 mg
Catalog # 1604665.
Supplier:   USP
Risperidone, Reference Standards
Catalog # 1604665.
Supplier:  USP
CAS Number:  
Risperidone, Reference Standards

Risperidone System Suitability Mixture, USP reference standard, Vial, 10 mg

CAS Number:  
Price
14 350,00 kr
Log in for availability
1 * 10 mg
Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications

Specifications

  • Pack type:
    Vial
  • Pk:
    10 mg
  • Standards:
    USP reference standard
  • Description:
    Risperidone System Suitability Mixture
  • UN No.:
    2811
  • Cat. No.:
    1604665.
  • Description:
    Risperidone

Specifications