Pyrocatechol ≥99.0% (by GC), Technical Grade
Glass bottle, 25 g
Catalog # TCIAP0317-25G
Supplier: TCI
Pyrocatechol ≥99.0% (by GC), Technical Grade
Catalog # TCIAP0317-25G
Supplier: TCI
CAS Number: 120-80-9
Jump to:
Product Details & Documents
Synonyms: o-Dihydroxybenzene, Catechol, 1,2-Dihydroxybenzene
- CAS Number:120-80-9
- Formula:C₆H₆O₂
- MDL Number:MFCD00002188
- Molecular Weight:110.11 g/mol
- Merck Index:14,07999
- Boiling Pt:246°C
- Melting Pt:103…105 °C
- Density:1,0548 g/cm³ (20 °C)
- Flash Pt:127 °C
- Storage Temperature:Ambient
- EINECS:204-427-5
- UN:2811
- ADR:6.1,III
Documents
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (GC):min. 99.0 %
- Melting point:104.0 to 107.0 deg-C
- Solubility in Water:transparency
Specifications
- Pack type:Glass bottle
- Pk:25 g



