Order Entry
Sweden
Orders LinkContactUsLinkComponent
 

Methyl violet 2B

Glass bottle, 25 g
Catalog # TCIAG0177-25G
Irritant
Health
Environment
Supplier:   TCI
Methyl violet 2B
Catalog # TCIAG0177-25G
Supplier:  TCI
CAS Number:  8004-87-3
Methyl violet 2B

Glass bottle, 25 g

CAS Number:  8004-87-3
Price
215,00 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Basic Violet 1, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]methylammonium chloride
  • CAS Number:8004-87-3
  • Formula:C₂₄H₂₈ClN₃
  • MDL Number:MFCD00012568
  • Molecular Weight:393.96 g/mol
  • Melting Pt:137 °C
  • Storage Temperature:Ambient
  • ADR:9,III
Specifications

Specification Test Results

  • Appearance:
    Green to Dark green powder to crystal
  • Lambda max.:
    583.0 to 587.0 nm (H2O)
  • Absorbance:
    min. 0.320 (2mg/L, H2O, 583.0 to 587.0 nm) (calcd.on dried substance)
  • abs-ratio (a/b):
    1.05 to 1.20 (E570/E600)
  • Drying loss:
    max. 10.0 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g

Specifications