Order Entry
Sweden
ContactUsLinkComponent
cHiPLC nanoflex system. For max. 3 cHiPLC jumper or column chips. Includes 10 port valve and temperature control for two chips. One cHiPLC column and two traps are included.Installation and training included if ordered as part of a system does not i 1 * 1 items
Catalog # 903-0607
Supplier:  EKSIGENT
cHiPLC nanoflex system. For max. 3 cHiPLC jumper or column chips. Includes 10 port valve and temperature control for two chips. One cHiPLC column and two traps are included.Installation and training included if ordered as part of a system does not i 1 * 1 items
Catalog # 903-0607
Supplier:  EKSIGENT
Frequently Bought Together