Order Entry
Sweden
ContactUsLinkComponent
ekspert microLC 200 binary gradient pump configured for 20-200 uL/min flow rate. Includes remotely mounted column oven, electrically -actuated low dispersion 6-port injection valve and mounting brackets for CTC six-plate (three drawer) cooled sample 1 * 1 items
Catalog # 903-0604
Supplier:  EKSIGENT
undefined
ekspert microLC 200 binary gradient pump configured for 20-200 uL/min flow rate. Includes remotely mounted column oven, electrically -actuated low dispersion 6-port injection valve and mounting brackets for CTC six-plate (three drawer) cooled sample 1 * 1 items
Catalog # 903-0604
Supplier:  EKSIGENT
Frequently Bought Together