Multi-parameter meters, handheld, Portavo® 907
Product Details & Documents
These versatile, portable instruments can measure pH, ORP, conductivity and dissolved oxygen (optical). Thanks to their robustness and large selection of suitable sensors, they are suitable for both laboratory and process applications. Waterproof according to IP 66/IP 67.
- Reinforced plastic enclosure for rough industrial applications
- Integrated cover protects the display and serves as laboratory stand
- High resolution colour TFT display is transreflective allowing the display to be read in bright sunlight
- Data log function can store up to 10000 values
- Digital Memosens® input, which allows signals to be digitised and transferred inductively i.e. via a non-contact connection
- Integrated sensor quiver (protective container for sensor) prevents sensor dry-out and damage
- Optical oxygen measurements possible
- Automatic status check of the device and sensor; evaluation of the sensor data end calibration intervals
- Memosens®: All information on sensor life are saved directly on the sensor (history of calibration, operation time, lifetime, sensor wear, SIP cycles…)
Portavo® 907 OXY Set includes the Portavo® 907 multi-parameter-meter and the digital probe SE 340 for optical oxygen measurements, with M12 8-pin plug according to DIN EN 50262.
Portavo® 907 LDO Set includes the Portavo® 907 multi-parameter-meter, the digital probe SE 340 for optical oxygen measurements, a carrying case and buffer solutions.
Portavo® 907 pH Set includes the Portavo® 907 multi-parameter-meter, the digital Memosens sensor SE 102 NMS, Memosens cable, carrying case and pH buffer solutions.
Portavo® 907 Multi Set includes the Portavo® 907 multi-parameter-meter, the digital Memosens pH sensor SE 102 NMS, Meosens conductivity sensor SE 615/1-MS, Memosens oxygen sensor SE 715/1-MS, Memosens cable, carrying case and pH buffer solutions.
Delivery information: Supplied with manual, 4× AA batteries, carrying strap, factory certificate, micro USB port for PC connection, USB cable (1,5 m) and Paraly® 112 PC software. All instruments, except for the sets, are delivered without an electrode, which needs to be ordered separately.
Portavo® 907 LDO Set includes the Portavo® 907 multi-parameter meter, SE340-MS digital probe for optical oxygen measurements, a carrying case and buffer solutions.
Portavo® 907 pH Set includes the Portavo® 907 multi-parameter meter, Memosens SE102-MS digital pH sensor, Memosens cable CA/MS-001XFA-L, carrying case and pH buffer solutions.
Portavo® 907 Multi Set includes the Portavo® 907 multi-parameter meter, Memosens SE102-MS digital pH sensor, Memosens SE615/1-MS conductivity sensor, Memosens SE715/1-MS oxygen sensor, Memosens cable CA/MS-001XFA-L, carrying case and pH buffer solutions.
Specifikationer
- Description:Portavo® 907 COND, without electrode, with analogue conductivity and digital Memosens input
- Seq:02
- Connections:1×multi contact for 2-/4-electrode sensors (analogue)
1×M8 socket, 4 pins, for Memosens lab cable
2×4 mm socket for separate temperature detector - Compliance:IP 66/IP 67
- Conductivity resolution:0,001 μS/cm (c <0,05 cm-1)
0,01 μS/cm (c = 0,05...0,2 cm-1)
0,1 μS/cm (c = 0,2...0,8 cm-1)
0,1 μS/cm (c >0,8 cm-1) - DO accuracy:Depending on Memosens electrode
- Data logging capacity:10000 values
- Power supply:4 alkaline AA batteries, Li-ion accumulator (loading via USB) available
- Resistivity range:00,00 - 99,99 MΩ cm
- Conductivity accuracy:<0,5% of value +0,4 μS
- W×D×H:127×30×155 mm
- Temperature range:NTC 30: −20...+120 °C
PT1000: −40...+250 °C
Memosens: −50...+250 °C - Salinity range:0,0 - 45,0 g/kg (0...+30 °C)
- mV range:±2000
- pH range:−2...+16
- DO concentration:0,00...0,99 / 1,00...20,00 mg/L
- Temperature accuracy:<0,5 K
- Weight:0,5 kg
- Cat. No.:665-0287
- pH accuracy:<±0,01
- Model:Portavo® 907 COND
- Conductivity range:4 electrode: 0,1 μS...1000 mS (0,05...500 mS/cm with electrode SE 204)
2 electrode: 0,1 μS...200 mS (0,01...200 μS/cm with electrode SE 202) - Outputs:Micro USB ports
- TDS range:0 - 1999 mg/l (10...40 °C)
- mV accuracy:<0,1% of value +0,3
- DO saturation:0,000...200,0 %
- Display:LCD, coloured
Accessories
Produktinformation
- Katalog #DescriptionTillgänglighetPrice
- 662-1826pH buffer set CaliMat, 4,00/7,00/9,00, ready-to-use; 250 ml eachLog in för att kolla tillgänglighet och ert pris1 * 1 SET - 640,00 kr
Produktinformation
- Katalog #DescriptionTillgänglighetPrice
- 662-3532PT1000 temperature probe, –10...+100 °C, Monel 2.4360, accuracy class A, acc. DIN IEC 751Log in för att kolla tillgänglighet och ert pris1 * 1 Items - 3 800,00 kr
Produktinformation
- Katalog #DescriptionTillgänglighetPrice
- 665-0300Adapter for connecting conductivity sensors with 2 banana plugs with instrumentLog in för att kolla tillgänglighet och ert pris1 * 1 Items - 1 550,00 kr
Produktinformation
- Katalog #DescriptionTillgänglighetPrice
- 665-0298KPG® tube for 4-electrode conductivity sensor ZU 6985, incl. O-ringLog in för att kolla tillgänglighet och ert pris1 * 1 Items - 1 275,00 kr
Produktinformation
- Katalog #DescriptionTillgänglighetPrice
- 665-0301Adapter for connecting 4 electrode sensor ZU 6985 with instrumentLog in för att kolla tillgänglighet och ert pris1 * 1 Items - 1 550,00 kr
Produktinformation
- Katalog #DescriptionTillgänglighetPrice
- 662-1599Robust field case (for meter, sensor, various small parts and user manual)Log in för att kolla tillgänglighet och ert pris1 * 1 Items - 2 200,00 kr
Produktinformation
- Katalog #DescriptionTillgänglighetPrice
- 663-2215PT1000 temperature probe, with tilted tip for puncture measurements in semi-solid substances; –10...+100 °C, Monel 2.4360, accuracy class A, acc. DIN IEC 751Log in för att kolla tillgänglighet och ert pris1 * 1 Items - 3 650,00 kr
Produktinformation
- Katalog #DescriptionTillgänglighetPrice
- 663-2112Conductivity standard, 12,88 mS/cm ±1 %, one ampoule for producing 1000 ml 0,1 mol/l NaCl solutionLog in för att kolla tillgänglighet och ert pris1 * 1 Items - 810,00 kr
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



