Lids for reaction vessels
flange NW 200, NS 45/40 (centre), 2×29/32 angled and 2×14/23 straight NS, 170 mm
Katalog # KGWAD4304
Leverantör: KGW
Gå till:
Product Details & Documents
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifikationer
- With:flange NW 200
- Seq:02
- Neck:NS 45/40 (centre)
- Side necks:2×29/32 angled and 2×14/23 straight NS
- Total height:170 mm