Order Entry
Sweden
ContactUsLinkComponent
PRODUCT FOR PROTEIN RELATE APPLICATION, PROTEIN STAINING SOLUTION, PROTEIN STAINING SOLUTION STAINS PROTEINS IN POLYACRYLAMIDE GEL A FTER SDS GEL ELECTROPHORESIS (DETECTION LIMIT >= 8 NG OF PROTEIN) WITH *COOMASSIER-G250. ALL PROCESSES INCLUDING WASH 1 * 500 mL
Catalog # ABNOTM0008
Supplier:  Abnova
PRODUCT FOR PROTEIN RELATE APPLICATION, PROTEIN STAINING SOLUTION, PROTEIN STAINING SOLUTION STAINS PROTEINS IN POLYACRYLAMIDE GEL A FTER SDS GEL ELECTROPHORESIS (DETECTION LIMIT >= 8 NG OF PROTEIN) WITH *COOMASSIER-G250. ALL PROCESSES INCLUDING WASH 1 * 500 mL
Catalog # ABNOTM0008
Supplier:  Abnova