Order Entry
Sweden
ContactUsLinkComponent
KIT, HISTONE H3 (TOTAL) QUANTIFICATION KIT (FLUOROMETRIC), HISTONE H3 (TOTAL) QUANTIFICATION KIT (FLUOROMETRIC) IS DESIGNED FOR QUAN TIFYING LEVELS OF HISTONE H3 PROTEINS INDEPENDENT OF ITS MODIFIED STATE AND CAN ALSO BE USED FOR NORMALIZING THE MODI 1 * 1 KIT
Catalog # ABNOKA0625
Leverantör:  Abnova
KIT, HISTONE H3 (TOTAL) QUANTIFICATION KIT (FLUOROMETRIC), HISTONE H3 (TOTAL) QUANTIFICATION KIT (FLUOROMETRIC) IS DESIGNED FOR QUAN TIFYING LEVELS OF HISTONE H3 PROTEINS INDEPENDENT OF ITS MODIFIED STATE AND CAN ALSO BE USED FOR NORMALIZING THE MODI 1 * 1 KIT
Catalog # ABNOKA0625
Leverantör:  Abnova
Relaterad information
Frequently Bought Together