Order Entry
Sweden
ContactUsLinkComponent
KIT, ADIPOQ (HUMAN) ELISA KIT, ADIPOQ (HUMAN) ELISA KIT IS DESIGNED FOR DETECTION OF ADIPOQ IN HUMAN URINE AND MURINE, ADIPONECTIN I N PLASMA, SERUM AND CELL CULTURE SUPERNATANTS. 1 * 1 KIT
Catalog # ABNOKA0978
Supplier:  Abnova
KIT, ADIPOQ (HUMAN) ELISA KIT, ADIPOQ (HUMAN) ELISA KIT IS DESIGNED FOR DETECTION OF ADIPOQ IN HUMAN URINE AND MURINE, ADIPONECTIN I N PLASMA, SERUM AND CELL CULTURE SUPERNATANTS. 1 * 1 KIT
Catalog # ABNOKA0978
Supplier:  Abnova
Related Information