Order Entry
Sweden
ContactUsLinkComponent
KIT, TFRC (HUMAN) ELISA KIT, TFRC (HUMAN) ELISA KIT IS A DOUBLE MONOCLONAL SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREM ENT OF HUMAN SOLUBLE TRANSFERRIN RECEPTOR IN SERUM, PLASMA AND TISSUE CULTURE MEDIUM. 1 * 1 KIT
  ABNOKA0039
 :  Abnova
KIT, TFRC (HUMAN) ELISA KIT, TFRC (HUMAN) ELISA KIT IS A DOUBLE MONOCLONAL SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREM ENT OF HUMAN SOLUBLE TRANSFERRIN RECEPTOR IN SERUM, PLASMA AND TISSUE CULTURE MEDIUM. 1 * 1 KIT
  ABNOKA0039
 :  Abnova
Frequently Bought Together