Order Entry
Sweden
ContactUsLinkComponent
KIT, LEP (HUMAN) ELISA KIT, LEP (HUMAN) CLINICAL RANGE ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT O F HUMAN LEPTIN IN SERUM, PLASMA AND TISSUE CULTURE MEDIUM. 1 * 1 KIT
Catalog # ABNOKA0025
Supplier:  Abnova
KIT, LEP (HUMAN) ELISA KIT, LEP (HUMAN) CLINICAL RANGE ELISA KIT IS A SANDWICH ENZYME IMMUNOASSAY FOR THE QUANTITATIVE MEASUREMENT O F HUMAN LEPTIN IN SERUM, PLASMA AND TISSUE CULTURE MEDIUM. 1 * 1 KIT
Catalog # ABNOKA0025
Supplier:  Abnova