Cover the reaction vessel with 8 connectors, outer diameter 184 mm, height 130 mm, DN 150, connctor 1 i 2 - treats GL 14 angle of 0 degrees from the vertical, PP cap and silicone gasket with a hole 10mm, connector 3 - oblique cut 29/32 angle of 10-15 1 * 1 items
Catalog # LENZ3.4508.CON
Supplier: Lenz Laborglas GmbH & CO.KG