Order Entry
Sweden
Orders LinkContactUsLinkComponent
COD standards
COD standard solution, 500 mg/l, 500 mL
  REGECOD500
 :   REAGECON
COD standards
  REGECOD500
 :  REAGECON
 :  
COD standards
COD standard solution, 500 mg/l, 500 mL
 :  
2 275,00 kr
1 * 500 mL
Ideal for use as control standards to verify that correct analysis for COD has taken place.
  • Concentration:
    500 mg/l
  • Pk:
    500 mL
  • Description:
    COD standard solution