Ribbons for label printer BBP™11
Thermal transfer printer ribbon, 4900 series, width 85 mm, BBP™11, 70 m, Black
BRDY804794
: Brady
- Colour:Black
- Description:Thermal transfer printer ribbon, 4900 series, width 85 mm
- Length:70 m
- Seq:09
- For:BBP™11



