Spore ampoules
Geobacillus stearothermophilus cell line 7953: spore ampoule106 (24 month shelf life) / sterilisation process: steam liquid cycles, 58 mm, 1 ml
Leverantör: CROSSTEX
Gå till:
Product Details & Documents
These spore ampoules are ideal for use in validation and routine monitoring of liquid steam sterilization cycles. Spore ampoules are manufactured using hermetically sealed Type I borosilicate glass containing an exclusively formulated Tryptic
Soy Broth (TSB) modified with pH indicator, and Geobacillus stearothermophilus spores. No activation is required; simply expose Spore Ampoules to sterilization process and incubate.
- Reduced incubation time of 48 hours
- Clear visible results as growth is evident by turbidity and/or a color shift of the media from purple to yellow
- Saves time by eliminating aseptic culturing required with traditional spore strips
- Unique packaging insert keeps ampoules in upright position and allows for quick reference to the number of units remaining
Delivery information: Each shipment is accompanied by a certificate of analysis and instructions for use
Caution: Storage under refrigerated conditions (2 to 8 °C) is required. Do not freeze spore ampoules.
Documents
Certificates
Sök efter LOT-nummerSpecifikationer
- Volume:1 ml
- Description:Geobacillus stearothermophilus cell line 7953: spore ampoule106 (24 month shelf life) / sterilisation process: steam liquid cycles
- Length:58 mm
- Seq:01



