Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DENSITY METER PORTABLE DMA35 1 * 1 items
Catalog # PAARDMA35
Supplier:
Anton Paar
Expand image
DENSITY METER PORTABLE DMA35 1 * 1 items
Catalog # PAARDMA35
Supplier:
Anton Paar