Order Entry
Sweden
ContactUsLinkComponent
Thermometer, ASTM, 57 C, white backed, -20+50:0,5C, mercury filled, immersion 57mm, c. 287x6-7mm long, durable pigment, government verified with Verification Certificate with 4 test points Until 10th October 2017 it is still allowed to use mercury 1 * 1 items
Catalog # 620-2593
Supplier:  Amarell
Thermometer, ASTM, 57 C, white backed, -20+50:0,5C, mercury filled, immersion 57mm, c. 287x6-7mm long, durable pigment, government verified with Verification Certificate with 4 test points Until 10th October 2017 it is still allowed to use mercury 1 * 1 items
Catalog # 620-2593
Supplier:  Amarell