Order Entry
Sweden
ContactUsLinkComponent
Thermometer, Landsberger, mercury filled, measuring range: +84+102/0.05C, enclosed scale, capillary lens front, colourless, total immersion, dimension: 300x9mm, richter-top finish with button, government verified, with verification certificate. Unt 1 * 1 items
Catalog # AMARN63841
Supplier:  Amarell
Thermometer, Landsberger, mercury filled, measuring range: +84+102/0.05C, enclosed scale, capillary lens front, colourless, total immersion, dimension: 300x9mm, richter-top finish with button, government verified, with verification certificate. Unt 1 * 1 items
Catalog # AMARN63841
Supplier:  Amarell