BUFFERTLÖSNING 25 C PH 4 1 * 500 mL
Catalog # FFCHFF63000500
Supplier: FF CHEMICALS
Frequently Bought Together
pH buffers, ready to use
Supplier: WTW
pH buffer solutions
Supplier: Mettler - Toledo
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
pH buffers, DuraCal
Supplier: HAMILTON BONADUZ
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Primary colour solutions, Reagent Ph. Eur.
Supplier: VWR Chemicals
Buffer solutions in sachets
Supplier: Mettler - Toledo
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals