Order Entry
Sweden
ContactUsLinkComponent
Base, petridish, material quartzglass, diameter 115mm, height 20mm. 1 * 1 items
Katalog # QUTW1173
Base, petridish, material quartzglass, diameter 115mm, height 20mm. 1 * 1 items
Katalog # QUTW1173