0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Base, petridish, material quartzglass, diameter 55mm, height 20mm. 1 * 1 items
Catalog # QUTW1171
Supplier:
QUARTZTECHNISCHE WERKSTATTEN
Expand image
Base, petridish, material quartzglass, diameter 55mm, height 20mm. 1 * 1 items
Catalog # QUTW1171
Supplier:
QUARTZTECHNISCHE WERKSTATTEN