Reagent sprayer for TLC plates, volume 250 mL, Bottle-Type 1 * 1 Each
Catalog # 58005.
Supplier: Merck
Reagent sprayer for TLC plates, volume 250 mL, Bottle-Type 1 * 1 Each
Catalog # 58005.
Supplier: Merck
Frequently Bought Together
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Air valves for safetycaps generation 2.0
Supplier: S.C.A.T.
Silica gel, Pore size: 60 Å 0.063-0.200 mm (70-230 mesh ASTM) for column chromatography, Millipore®
Supplier: MERCK PRODUCTION CHEMICALS
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
Exhaust filters, V3.0
Supplier: S.C.A.T.
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Short Thread Vials ND9
Supplier: VWR Collection
VWR®, Crimp Neck Vials ND11
Supplier: VWR Collection
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Crimp Caps for Crimp Neck Vials ND11
Supplier: VWR Collection
VWR®, Screw Neck Vials ND13
Supplier: VWR Collection
Ethyl levulinate 98%
Supplier: Thermo Fisher Scientific