Order Entry
Sweden
ContactUsLinkComponent
Metal Packed GC Column (General Configuration), phase 10% Carbowax 20M, matrix 80/100 Chromosorb W AW support, L x O.D. x I.D. 6.0 f t (1.8 m) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12212.
Supplier:  Merck
Metal Packed GC Column (General Configuration), phase 10% Carbowax 20M, matrix 80/100 Chromosorb W AW support, L x O.D. x I.D. 6.0 f t (1.8 m) x 1/8 in. x 2.1 mm 1 * 1 Each
Catalog # 12212.
Supplier:  Merck