0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Coated GC Packing, phase 6.6% Carbowax 20M, matrix 80/120 Carbopack B AW support, bottle of 15 g 1 * 15 g
Catalog # 11814.
Supplier:
Merck
Expand image
Coated GC Packing, phase 6.6% Carbowax 20M, matrix 80/120 Carbopack B AW support, bottle of 15 g 1 * 15 g
Catalog # 11814.
Supplier:
Merck