Order Entry
Sweden
ContactUsLinkComponent
Freezer chest,glk30 -10 to -60 degree celsius, volume 300litres, laboratory use, electronic thermostat, climate class 4 (en 441), wi th loc k, defrost drain, acoustic alarm, length 1305mm, depth 655mm, height 865mm, insulation thickness 100mm. 1 * 1 items
  LIEBGLK30
 :  Liebherr
Freezer chest,glk30 -10 to -60 degree celsius, volume 300litres, laboratory use, electronic thermostat, climate class 4 (en 441), wi th loc k, defrost drain, acoustic alarm, length 1305mm, depth 655mm, height 865mm, insulation thickness 100mm. 1 * 1 items
  LIEBGLK30
 :  Liebherr