Order Entry
Sweden
Orders LinkContactUsLinkComponent
Shelving systems, direct wall mounts
Shelving systems, direct wall mounts
Shelf, single, 0,9 kg
Catalog # METO1WD18S
Supplier:   Metro International
Shelving systems, direct wall mounts
Catalog # METO1WD18S
Shelving systems, direct wall mounts
Shelving systems, direct wall mounts
Shelf, single, 0,9 kg
Price
1 675,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Direct wall mount brackets allow the installation of wire or solid shelves directly onto a wall surface. Each brackets consists of one shelf support and mounting plate, with two shelf collar caps. Use single support at shelf ends and double support for adjoining shelves.
Specifications
  • Description:
    Shelf, single
  • Seq:
    03
  • Weight:
    0,9 kg