Order Entry
Sweden
Orders LinkContactUsLinkComponent
Formalin 10% in aqueous phosphate buffer (pH 7.0 ± 0.2)

Formalin 10% in aqueous phosphate buffer (pH 7.0 ± 0.2)

Glass bottle, 150 mL
Catalog # FFCHFF8370150
Health
Irritant
Supplier:   FF CHEMICALS
Formalin 10% in aqueous phosphate buffer (pH 7.0 ± 0.2)
Catalog # FFCHFF8370150
Supplier:  FF CHEMICALS
CAS Number:  50-00-0
Formalin 10% in aqueous phosphate buffer (pH 7.0 ± 0.2)

Glass bottle, 150 mL

CAS Number:  50-00-0
Price
Inquire For Price
Log in for availability
1 * 150 mL
Product Details & Documents

(= 4% Formaldehyde solution)

Synonyms:  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  • CAS Number:50-00-0
  • Formula:CH₂O
  • MDL Number:MFCD00003274
  • Molecular Weight:30.03 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-001-8
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    150 mL

Specifications