Order Entry
Sweden
Orders LinkContactUsLinkComponent
Formalin 10% in aqueous phosphate buffer (pH 7.0 ± 0.2)
3 L
  FFCHFF22023000
Formalin 10% in aqueous phosphate buffer (pH 7.0 ± 0.2)
  FFCHFF22023000
 :  50-00-0
Formalin 10% in aqueous phosphate buffer (pH 7.0 ± 0.2)
3 L
 :  50-00-0
1 * 3 L
(= 4% Formaldehyde solution)
 :  Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
  •  :50-00-0
  •  :CH₂O
  •  :MFCD00003274
  •  :30.03 g/mol
  •  :Ambient
  •  :200-001-8
  • Pk:
    3 L