Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
BUFFER SOLUTION PH4 1 * 500 mL
Catalog # FFCHFF1640500
Supplier:
FF CHEMICALS
Certificates
Expand image
BUFFER SOLUTION PH4 1 * 500 mL
Catalog # FFCHFF1640500
Supplier:
FF CHEMICALS
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation