Order Entry
Sweden
Orders LinkContactUsLinkComponent
Pipette tips
In box, 10 ml, -, 35 tips/box
Catalog # VIST9050
Pipette tips
Catalog # VIST9050
Pipette tips
Pipette tips
In box, 10 ml, -, 35 tips/box
Price
470,00 kr
Log in for availability
1 * 35 Items
Product Details & Documents
These tips to maximise the exceptional performance and benefits of ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
Specifications
  • Packed:
    35 tips/box
  • Sterile:
    -
  • Volume:
    10 ml
  • Seq:
    24
  • Tip style:
    In box