Pipette tips
VistaTip™, pyrgen-free, 1250 µl, +, Individually wrapped
VIST2028
These tips to maximise the exceptional performance and benefits of ovation and MLA pipettes. All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time.
- Packed:Individually wrapped
- Sterile:+
- Volume:1250 µl
- Seq:10
- Tip style:VistaTip™, pyrgen-free