0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Tip pipette 200µl HLT 200 low retention clear beveled orifice graduations at 10-50-100µl rack non-sterile 1 * 960 items
Catalog # MLBP3932-05
Supplier:
Thermo Fisher Scientific
Expand image
Tip pipette 200µl HLT 200 low retention clear beveled orifice graduations at 10-50-100µl rack non-sterile 1 * 960 items
Catalog # MLBP3932-05
Supplier:
Thermo Fisher Scientific